5-(3,4-Dimethoxyphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35966-500MG 35966-1G 35966-5G 35966-25G
77325-978EA 134.93 USD
77325-978 77344-454 77362-284 77385-256
5-(3,4-Dimethoxyphenyl)oxazole ≥97%
5-(3,4-Dimethoxyphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD32631885
CAS Number: 73663-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR