5-(3-Fluoro-4-methoxyphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35965-500MG 35965-1G 35965-5G 35965-25G
77325-976EA 119.48 USD
77325-976 77343-628 77362-282 77373-888
5-(3-Fluoro-4-methoxyphenyl)oxazole ≥97%
5-(3-Fluoro-4-methoxyphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₂
MW: 193.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631884
CAS Number: 1904415-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR