5-(2-Fluoro-6-(trifluoromethyl)phenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35964-500MG 35964-1G 35964-5G 35964-25G
77325-974EA 134.93 USD
77325-974 77344-452 77362-280 77385-254
5-(2-Fluoro-6-(trifluoromethyl)phenyl)oxazole ≥97%
5-(2-Fluoro-6-(trifluoromethyl)phenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₅F₄NO
MW: 231.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631883
CAS Number: 2364585-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR