5-(2,3-Difluoro-6-methoxyphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35963-500MG 35963-1G 35963-5G
77325-972EA 317.24 USD
77325-972 77349-674 77373-886
5-(2,3-Difluoro-6-methoxyphenyl)oxazole ≥97%
5-(2,3-Difluoro-6-methoxyphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₇F₂NO₂
MW: 211.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631882
CAS Number: 2364585-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR