5-(6-Bromo-2,3-dichlorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35961-500MG 35961-1G 35961-5G
77339-054EA 547.96 USD
77339-054 77349-670 77373-884
5-(6-Bromo-2,3-dichlorophenyl)oxazole ≥97%
5-(6-Bromo-2,3-dichlorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄BrCl₂NO
Storage Temperature: Ambient
MDL Number: MFCD32631880
CAS Number: 2364585-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR