5-(2,3-Dichloro-6-fluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35957-500MG 35957-1G 35957-5G
77325-964EA 311.06 USD
77325-964 77349-666 77373-882
5-(2,3-Dichloro-6-fluorophenyl)oxazole ≥97%
5-(2,3-Dichloro-6-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄Cl₂FNO
Storage Temperature: Ambient
MDL Number: MFCD32631877
CAS Number: 2364584-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR