5-(3-Bromo-2-chloro-6-fluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35956-500MG 35956-1G 35956-5G
77325-962EA 311.06 USD
77325-962 77349-664 77373-880
5-(3-Bromo-2-chloro-6-fluorophenyl)oxazole ≥97%
5-(3-Bromo-2-chloro-6-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄BrClFNO
MW: 276.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631876
CAS Number: 2364584-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR