5-(4-(benzyloxy)-3-fluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35955-500MG 35955-1G 35955-25G 35955-5G
77325-960EA 292.52 USD
77325-960 77349-662 77385-246 77373-878
5-(4-(benzyloxy)-3-fluorophenyl)oxazole ≥97%
5-(4-(benzyloxy)-3-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₆H₁₂FNO₂
MW: 269.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631875
CAS Number: 2328072-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR