5-(4-Bromo-2,6-difluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35952-500MG 35952-1G 35952-5G 35952-25G
77325-956EA 156.56 USD
77325-956 77345-616 77362-272 77385-244
5-(4-Bromo-2,6-difluorophenyl)oxazole ≥97%
5-(4-Bromo-2,6-difluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄BrF₂NO
MW: 260.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631872
CAS Number: 1400706-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR