5-(2-Bromo-6-fluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35949-500MG 35949-1G 35949-5G 35949-25G
77325-952EA 129.78 USD
77325-952 77344-304 77362-268 77373-870
5-(2-Bromo-6-fluorophenyl)oxazole ≥97%
5-(2-Bromo-6-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₅BrFNO
MW: 242.045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631869
CAS Number: 2024254-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR