5-(2-Bromo-4,6-difluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35946-500MG 35946-1G 35946-5G
77339-566EA 597.4 USD
77339-566 77349-654 77373-868
5-(2-Bromo-4,6-difluorophenyl)oxazole ≥97%
5-(2-Bromo-4,6-difluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄BrF₂NO
Storage Temperature: Ambient
MDL Number: MFCD32631866
CAS Number: 2364584-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR