5-(4-Bromo-2-fluoro-5-methylphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35943-500MG 35943-1G 35943-5G
77338-128EA 466.59 USD
77338-128 77349-652 77373-866
5-(4-Bromo-2-fluoro-5-methylphenyl)oxazole ≥97%
5-(4-Bromo-2-fluoro-5-methylphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₇BrFNO
MW: 256.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631864
CAS Number: 2364585-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR