5-(2,4-bis(trifluoromethyl)phenyl)oxazole ≥95%

Supplier: AOBChem USA

Warning

35940-500MG 35940-1G 35940-5G 35940-25G
77325-946EA 182.31 USD
77325-946 77346-930 77362-262 77385-236
5-(2,4-bis(trifluoromethyl)phenyl)oxazole ≥95%
5-(2,4-bis(trifluoromethyl)phenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₁H₅F₆NO
MW: 281.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631861
CAS Number: 2364585-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR