5-(2,6-Dichloro-4-methylphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35938-500MG 35938-1G 35938-5G
77338-304EA 473.8 USD
77338-304 77349-648 77373-862
5-(2,6-Dichloro-4-methylphenyl)oxazole ≥97%
5-(2,6-Dichloro-4-methylphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₇Cl₂NO
MW: 228.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631859
CAS Number: 2364585-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR