5-(2,4,6-trichlorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35937-500MG 35937-1G 35937-5G 35937-25G
77325-942EA 238.96 USD
77325-942 77349-646 77362-260 77385-234
5-(2,4,6-trichlorophenyl)oxazole ≥97%
5-(2,4,6-trichlorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄Cl₃NO
MW: 248.493 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631858
CAS Number: 2364585-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR