5-(4-Bromo-2-methoxyphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35934-500MG 35934-1G 35934-5G 35934-25G
77325-940EA 259.56 USD
77325-940 77349-642 77362-258 77385-232
5-(4-Bromo-2-methoxyphenyl)oxazole ≥97%
5-(4-Bromo-2-methoxyphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631855
CAS Number: 1091618-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR