5-(2,6-Dibromophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35933-500MG 35933-1G 35933-5G 35933-25G
77325-938EA 259.56 USD
77325-938 77349-640 77362-256 77385-230
5-(2,6-Dibromophenyl)oxazole ≥97%
5-(2,6-Dibromophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₅Br₂NO
MW: 302.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631854
CAS Number: 2364585-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR