5-(2-Bromo-5-methylphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35932-500MG 35932-1G 35932-5G 35932-25G
77325-936EA 259.56 USD
77325-936 77349-638 77362-254 77385-228
5-(2-Bromo-5-methylphenyl)oxazole ≥97%
5-(2-Bromo-5-methylphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MW: 238.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631853
CAS Number: 2028261-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR