5-(4-(benzyloxy)-3,5-dibromophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35931-500MG 35931-1G 35931-5G
77338-302EA 473.8 USD
77338-302 77349-636 77373-858
5-(4-(benzyloxy)-3,5-dibromophenyl)oxazole ≥97%
5-(4-(benzyloxy)-3,5-dibromophenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₆H₁₁Br₂NO₂
MW: 409.072 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631852
CAS Number: 2364584-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR