5-(2,4,6-Tribromophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35929-500MG 35929-1G 35929-5G
77325-934EA 311.06 USD
77325-934 77349-634 77373-856
5-(2,4,6-Tribromophenyl)oxazole ≥97%
5-(2,4,6-Tribromophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄Br₃NO
Storage Temperature: Ambient
MDL Number: MFCD32631850
CAS Number: 2364584-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR