5-(2-Bromo-4-methylphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35928-500MG 35928-1G 35928-5G
77325-932EA 320.33 USD
77325-932 77349-632 77373-854
5-(2-Bromo-4-methylphenyl)oxazole ≥97%
5-(2-Bromo-4-methylphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MW: 238.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27933771
CAS Number: 938461-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR