5-(4-Bromo-2,6-dichlorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35927-500MG 35927-1G 35927-5G
77325-930EA 336.81 USD
77325-930 77349-630 77373-852
5-(4-Bromo-2,6-dichlorophenyl)oxazole ≥97%
5-(4-Bromo-2,6-dichlorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄BrCl₂NO
MW: 292.944 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631849
CAS Number: 2379321-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR