5-(4-Chloro-2-fluorophenyl)oxazole ≥95%

Supplier: AOBChem USA

Warning

35925-500MG 35925-1G 35925-5G 35925-25G
77325-928EA 259.56 USD
77325-928 77349-628 77362-252 77385-226
5-(4-Chloro-2-fluorophenyl)oxazole ≥95%
5-(4-Chloro-2-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₅ClFNO
Storage Temperature: Ambient
MDL Number: MFCD32631847
CAS Number: 2020663-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR