5-(3,6-Dibromo-2-fluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35924-500MG 35924-1G 35924-5G 35924-25G
77325-926EA 233.81 USD
77325-926 77349-626 77362-250 77385-224
5-(3,6-Dibromo-2-fluorophenyl)oxazole ≥97%
5-(3,6-Dibromo-2-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄Br₂FNO
MW: 320.941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631846
CAS Number: 2379321-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR