5-(4-Bromo-2,3-dimethylphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35923-500MG 35923-1G 35923-5G
77325-924EA 336.81 USD
77325-924 77349-624 77373-850
5-(4-Bromo-2,3-dimethylphenyl)oxazole ≥97%
5-(4-Bromo-2,3-dimethylphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO
MW: 252.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631845
CAS Number: 2364585-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR