Methyl 5-methyl-1H-indazole-4-carboxylate ≥95%

Supplier: AOBChem USA

Warning

35898-500MG 35898-1G 35898-5G
77339-108EA 551.05 USD
77339-108 77349-622 77373-848
Methyl 5-methyl-1H-indazole-4-carboxylate ≥95%

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD23705404
CAS Number: 1427369-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR