tert-Butyl 2-fluoro-5-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

35890-1G 35890-5G 35890-25G
77341-086EA 70.04 USD
77341-086 77362-248 77373-846
tert-Butyl 2-fluoro-5-iodobenzoate ≥97%
tert-Butyl 2-fluoro-5-iodobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂FIO₂
Storage Temperature: Ambient
MDL Number: MFCD32631844
CAS Number: 1382948-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR