tert-Butyl 3,4-dibromo-5-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

35866-500MG 35866-1G 35866-5G
77336-590EA 389.34 USD
77336-590 77349-620 77373-840
tert-Butyl 3,4-dibromo-5-methylbenzoate ≥95%
tert-Butyl 3,4-dibromo-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄Br₂O₂
MW: 350.046 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631839
CAS Number: 2364585-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR