4-Bromo-N-tert-butyl-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

35856-500MG 35856-1G 35856-5G 35856-25G
77325-918EA 110.21 USD
77325-918 77343-260 77362-242 77373-836
4-Bromo-N-tert-butyl-3-methylbenzamide ≥97%
4-Bromo-N-tert-butyl-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD27001836
CAS Number: 149105-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR