4-Bromo-N-cyclopropyl-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

35849-500MG 35849-1G 35849-5G 35849-25G
77325-916EA 99.91 USD
77325-916 77342-776 77362-238 77373-832
4-Bromo-N-cyclopropyl-3-methylbenzamide ≥97%
4-Bromo-N-cyclopropyl-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrNO
Storage Temperature: Ambient
MDL Number: MFCD26802224
CAS Number: 1564656-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR