5-(4-Bromo-2-ethylphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35831-500MG 35831-1G 35831-5G
77339-288EA 572.68 USD
77339-288 77349-616 77373-830
5-(4-Bromo-2-ethylphenyl)oxazole ≥97%
5-(4-Bromo-2-ethylphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO
MW: 252.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631830
CAS Number: 2364585-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR