5-(2-Bromo-4-(trifluoromethoxy)phenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35830-500MG 35830-1G 35830-5G
77338-300EA 473.8 USD
77338-300 77349-614 77373-828
5-(2-Bromo-4-(trifluoromethoxy)phenyl)oxazole ≥97%
5-(2-Bromo-4-(trifluoromethoxy)phenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₅BrF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD32631829
CAS Number: 2364584-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR