5-(2-Bromo-4-(trifluoromethyl)phenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35816-500MG 35816-1G 35816-5G
77325-914EA 336.81 USD
77325-914 77349-612 77373-826
5-(2-Bromo-4-(trifluoromethyl)phenyl)oxazole ≥97%
5-(2-Bromo-4-(trifluoromethyl)phenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₅BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD32631822
CAS Number: 2364585-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR