(2-Bromo-5-(methoxycarbonyl)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

35809-500MG 35809-1G 35809-5G
77338-282EA 471.74 USD
77338-282 77349-610 77373-824
(2-Bromo-5-(methoxycarbonyl)phenyl)boronic acid ≥95%
(2-Bromo-5-(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BBrO₄
MW: 258.862 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24449009
CAS Number: 2225178-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR