4-Bromo-3-methylbenzoic acid benzyl ester ≥95%

Supplier: AOBChem USA

Warning

35807-500MG 35807-1G 35807-5G 35807-25G
77325-912EA 213.21 USD
77325-912 77349-608 77362-236 77385-222
4-Bromo-3-methylbenzoic acid benzyl ester ≥95%
4-Bromo-3-methylbenzoic acid benzyl ester

Precaución: for Research Use Only

Formula: C₁₅H₁₃BrO₂
MW: 305.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30536300
CAS Number: 591775-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR