5-[3-(Propan-2-yl)phenyl]-1,3-oxazole ≥97%

Supplier: AOBChem USA

Warning

35801-500MG 35801-1G 35801-5G 35801-25G
77325-910EA 259.56 USD
77325-910 77349-606 77362-234 77385-220
5-[3-(Propan-2-yl)phenyl]-1,3-oxazole ≥97%
5-[3-(Propan-2-yl)phenyl]-1,3-oxazole

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD32631820
CAS Number: 2364585-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR