1-(2-(methoxymethyl)phenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

35776-500MG 35776-1G 35776-5G 35776-25G
77325-902EA 264.71 USD
77325-902 77349-598 77362-226 77385-212
1-(2-(methoxymethyl)phenyl)ethanol ≥97%
1-(2-(methoxymethyl)phenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD21267543
CAS Number: 1087390-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR