5-(4-Bromo-2-methylphenyl)-1,3-oxazole ≥97%

Supplier: AOBChem USA

Warning

35733-500MG 35733-1G 35733-5G 35733-25G
77325-894EA 208.06 USD
77325-894 77349-592 77362-220 77385-208
5-(4-Bromo-2-methylphenyl)-1,3-oxazole ≥97%
5-(4-Bromo-2-methylphenyl)-1,3-oxazole

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD31725130
CAS Number: 501944-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR