5-(4-Bromo-3-chlorophenyl)-1,3-oxazole ≥97%

Supplier: AOBChem USA

Warning

35728-500MG 35728-1G 35728-5G
77325-890EA 311.06 USD
77325-890 77349-588 77373-814
5-(4-Bromo-3-chlorophenyl)-1,3-oxazole ≥97%
5-(4-Bromo-3-chlorophenyl)-1,3-oxazole

Precaución: for Research Use Only

Formula: C₉H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD32215242
CAS Number: 1997950-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR