5-(3-Bromo-4-chlorophenyl)-1,3-oxazole ≥97%

Supplier: AOBChem USA

Warning

35727-500MG 35727-1G 35727-5G
77325-888EA 311.06 USD
77325-888 77349-586 77373-812
5-(3-Bromo-4-chlorophenyl)-1,3-oxazole ≥97%
5-(3-Bromo-4-chlorophenyl)-1,3-oxazole

Precaución: for Research Use Only

Formula: C₉H₅BrClNO
MW: 258.499 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32215243
CAS Number: 2002122-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR