5-(5-Bezyloxy-2-bromophenyl)-1,3-oxazole ≥97%

Supplier: AOBChem USA

Warning

35720-500MG 35720-1G 35720-25G 35720-5G
77325-886EA 292.52 USD
77325-886 77349-584 77385-206 77373-810
5-(5-Bezyloxy-2-bromophenyl)-1,3-oxazole ≥97%
5-(5-Bezyloxy-2-bromophenyl)-1,3-oxazole

Precaución: for Research Use Only

Formula: C₁₆H₁₂BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD32215235
CAS Number: 2364585-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR