5-(2,6-Difluorophenyl)-1,3-oxazole ≥97%

Supplier: AOBChem USA

Warning

35713-500MG 35713-1G 35713-5G 35713-25G
77325-882EA 259.56 USD
77325-882 77349-580 77362-216 77385-202
5-(2,6-Difluorophenyl)-1,3-oxazole ≥97%
5-(2,6-Difluorophenyl)-1,3-oxazole

Precaución: for Research Use Only

Formula: C₉H₅F₂NO
Storage Temperature: Ambient
MDL Number: MFCD32215258
CAS Number: 2001301-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR