5-(5-Bromothiophen-2-yl)-1,3-oxazole ≥97%

Supplier: AOBChem USA

Warning

35711-500MG 35711-1G 35711-5G 35711-25G
77325-880EA 244.11 USD
77325-880 77349-576 77362-214 77385-200
5-(5-Bromothiophen-2-yl)-1,3-oxazole ≥97%
5-(5-Bromothiophen-2-yl)-1,3-oxazole

Precaución: for Research Use Only

Formula: C₇H₄BrNOS
Storage Temperature: Ambient
MDL Number: MFCD02682014
CAS Number: 321309-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR