5-[3-Bromo-5-(trifluoromethyl)phenyl]-oxazole ≥97%

Supplier: AOBChem USA

Warning

35710-500MG 35710-1G 35710-5G
77335-672EA 363.59 USD
77335-672 77349-574 77373-806
5-[3-Bromo-5-(trifluoromethyl)phenyl]-oxazole ≥97%
5-[3-Bromo-5-(trifluoromethyl)phenyl]-oxazole

Precaución: for Research Use Only

Formula: C₁₀H₅BrF₃NO
MW: 292.052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32215260
CAS Number: 1630747-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR