Isopropyl 4-bromobenzoate ≥95%

Supplier: AOBChem USA

Warning

35697-500MG 35697-1G 35697-5G 35697-25G
77325-878EA 199.82 USD
77325-878 77349-570 77362-212 77385-198
Isopropyl 4-bromobenzoate ≥95%
Isopropyl 4-bromobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD21101314
CAS Number: 59247-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR