5-Bromo-2,3-difluoro-4-ethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

35611-500MG 35611-1G 35611-5G
77335-468EA 355.35 USD
77335-468 77349-568 77373-802
5-Bromo-2,3-difluoro-4-ethoxyphenol ≥97%
5-Bromo-2,3-difluoro-4-ethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O₂
MW: 253.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32215264
CAS Number: 2271443-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR