3,4-Difluoro-2-ethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

35608-500MG 35608-1G 35608-5G 35608-25G
77325-872EA 254.41 USD
77325-872 77349-566 77362-206 77385-196
3,4-Difluoro-2-ethoxyphenol ≥97%
3,4-Difluoro-2-ethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD32215265
CAS Number: 2271443-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR