3,4-Difluoro-2-ethoxyphenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
35607-500MG
35607-1G
35607-5G
35607-25G
77325-870EA
218.36
USD
77325-870
77349-564
77362-204
77385-194
3,4-Difluoro-2-ethoxyphenylboronic acid pinacol ester ≥97%
3,4-Difluoro-2-ethoxyphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₁₉BF₂O₃ MW: 284.107 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32200306 CAS Number: 2877757-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...