[2-Chloro-3-fluoro-4-(methylsulfanyl)phenyl]methanol ≥95%

Supplier: AOBChem USA

Warning

35604-500MG 35604-1G 35604-5G
77325-864EA 316.21 USD
77325-864 77349-560 77373-794
[2-Chloro-3-fluoro-4-(methylsulfanyl)phenyl]methanol ≥95%
[2-Chloro-3-fluoro-4-(methylsulfanyl)phenyl]methanol

Precaución: for Research Use Only

Formula: C₈H₈ClFOS
MW: 206.665 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32215266
CAS Number: 2271442-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR