4-Nitro-1-(methoxymethoxy)biphenyl ≥95%

Supplier: AOBChem USA

Warning

35600-500MG 35600-1G 35600-5G 35600-25G
77325-862EA 208.06 USD
77325-862 77349-558 77362-200 77385-190
4-Nitro-1-(methoxymethoxy)biphenyl ≥95%
4-Nitro-1-(methoxymethoxy)biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD32631813
CAS Number: 2271442-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR